missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iodide, puriss., Honeywell Fluka™
Puriss.,≥99%
$885.28 - $885.28
Chemical Identifiers
| CAS | 12027-06-4 |
|---|---|
| Molecular Formula | H4IN |
| Molecular Weight (g/mol) | 144.94 |
| MDL Number | MFCD00011424 |
| InChI Key | UKFWSNCTAHXBQN-UHFFFAOYSA-N |
| Synonym | ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i |
| PubChem CID | 25487 |
| ChEBI | CHEBI:63040 |
| SMILES | [NH4+].[I-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 12027-06-4 | |
| 144.94 | |
| UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
| 25487 | |
| [NH4+].[I-] |
| H4IN | |
| MFCD00011424 | |
| ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i | |
| CHEBI:63040 |
Specifications
| 12027-06-4 | |
| H4IN | |
| MFCD00011424 | |
| ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i | |
| [NH4+].[I-] | |
| 144.94 | |
| CHEBI:63040 | |
| ≥99% | |
| ≤0.1% (as SO4) | |
| Ammonium Iodide |
| 1 kg | |
| NH4I | |
| NONH for all modes of transport | |
| UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
| azanium;iodide | |
| 25487 | |
| 144.94g/mol | |
| puriss. | |
| 1mmHg (210.9°C) |
Safety and Handling
P261-P305 + P351 + P338
H315-H319-H335
EINECSNumber : 234-717-7