Learn More
Ammonium iodide, ACS reagent
CAS: 12027-06-4 | H4IN | 144.94 g/mol
$104.43 - $104.43
Chemical Identifiers
| CAS | 12027-06-4 |
|---|---|
| Molecular Formula | H4IN |
| Molecular Weight (g/mol) | 144.94 |
| MDL Number | MFCD00011424 |
| InChI Key | UKFWSNCTAHXBQN-UHFFFAOYSA-N |
| Synonym | ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i |
| PubChem CID | 25487 |
| ChEBI | CHEBI:63040 |
| IUPAC Name | azanium;iodide |
| SMILES | [NH4+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC193210500
|
Thermo Scientific Chemicals
193210500 |
50 g | Glass bottle |
Each for $104.43
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12027-06-4 | |
| 144.94 | |
| UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
| 25487 | |
| azanium;iodide |
| H4IN | |
| MFCD00011424 | |
| ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i | |
| CHEBI:63040 | |
| [NH4+].[I-] |
Specifications
| 551.0°C | |
| 50 g | |
| NH4I | |
| 15,526 | |
| Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C), 270g/L alcohol - 667g/L glycerol, 400g/L methanol | |
| [NH4+].[I-] | |
| 144.94 | |
| CHEBI:63040 | |
| ACS Reagent | |
| 0.05% max | |
| 2.5100g/mL |
| 12027-06-4 | |
| H4IN | |
| MFCD00011424 | |
| ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i | |
| UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
| azanium;iodide | |
| 25487 | |
| 144.94 | |
| Glass bottle | |
| 2.51 | |
| Ammonium iodide |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
EINECSNumber : 234-717-7
RUO – Research Use Only