missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iodide, (ACS Reagent Grade), MP Biomedicals

Catalog No. p-3547006
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15249680 100 g
ICN15249690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15249680 Supplier MP Biomedicals Inc Supplier No. 0215249680
Only null left

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name ammonium iodide
SMILES [NH4+].[I-]

Specifications

Quantity 100 g
Formula Weight 144.9
Grade ACS Reagent
Chemical Name or Material Ammonium Iodide
Recommended Storage Room Temperature