missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iodide, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor A1205500GM
Specifications
| 12027-06-4 | |
| 500 g | |
| UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
| ammonium iodide | |
| 99% | |
| Amber Glass Bottle |
| 1 | |
| H4IN | |
| [NH4+].[I-] | |
| 144.94 | |
| ACS | |
| 0.0005 |
Chemical Identifiers
| 12027-06-4 | |
| 144.94 | |
| ammonium iodide |
| H4IN | |
| UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
| [NH4+].[I-] |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.