missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 99+%, for analysis

Catalog No. p-45518
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206461000 100 g Glass bottle
AC206465000 500 g Glass bottle
AC206460025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC206461000 Supplier Thermo Scientific Chemicals Supplier No. 206461000
Only null left

CAS: 12027-06-4 | H4IN | 144.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551.0°C
Color White to Yellow
pH 4.5 to 6.5 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula NH4I
Merck Index 15, 526
Solubility Information Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C), 270g/L alcohol - 667g/L glycerol, 400g/L methanol
Formula Weight 144.94
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Specific Gravity 2.51
Density 2.5100g/mL
Chemical Name or Material Ammonium iodide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 234-717-7
TSCA TSCA

RUO – Research Use Only