missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 99.995% (metals basis)

Catalog No. AA4025506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4025506 5 g
AA4025514 25 g
AA4025522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4025506 Supplier Thermo Scientific Chemicals Supplier No. 04025506

Light Sensitive and Hygroscopic | CAS: 12027-06-4 | H4IN | 144.94 g/mol

It is used in photographic chemicals and some medications. Ammonium iodide and hydrogen peroxide plays an essential role for the eco-friendly oxyiodination of aromatic compounds. Ammonium Iodide and DMF is also useful for the copper-mediated selective cyanation reaction of Indoles and 2-Phenylpyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551°C (sublimation)
Color White to Yellow
Quantity 5 g
Linear Formula NH4I
Merck Index 14,529
Solubility Information Soluble in water and ethanol.
Formula Weight 144.94
Percent Purity 99.995%
Odor Odorless
Refractive Index 1.7031
Sensitivity Air and light sensitive; Hygroscopic
Density 2.514
Chemical Name or Material Ammonium iodide, (metals basis)
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-717-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only