missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 98+%

Catalog No. AAA1249822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249822 100 g
AAA1249836 500 g
AAA124980E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1249822 Supplier Thermo Scientific Chemicals Supplier No. A1249822

CAS: 12027-06-4 | H4IN | 144.94 g/mol

Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.

Notes
Store at 4°C. Hande under dry protective gas.

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551°C (sublimation)
Quantity 100 g
Merck Index 14,529
Solubility Information Soluble in water (1770mg/ml at 25°C),methanol (400mg/ml),glycerol (667mg/ml),alcohol (270mg/ml),and ethanol.
Formula Weight 144.94
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.7031
Sensitivity Air and light sensitive; Hygroscopic
Density 2.514
Chemical Name or Material Ammonium iodide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-717-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only