missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, TraceGrade, 20-22%, Spectrum™ Chemical

Catalog No. p-7164955
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608685 500 mL Poly Bottle
18608684 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608685 Supplier Spectrum Chemical Mfg Cor Supplier No. A1069500MLPL
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7664-41-7
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 1
Quantity 500 mL
Percent Purity 20 to 22%
Grade High Purity Solvent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.