missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Fluka, Honeywell
$2223.08 - $2223.08
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| MDL Number | MFCD00066650 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Ammonia aqueous, Ammonia water |
| IUPAC Name | water |
| SMILES | O |
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
| H2O | |
| MFCD00066650 | |
| Ammonia aqueous, Ammonia water | |
| O |
Specifications
| 24°C | |
| ≥70.00,≥20.00 | |
| H2O | |
| 3587154 | |
| Ammonia aqueous, Ammonia water | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Trace Analysis | |
| Liquid |
| 7732-18-5,1336-21-6 | |
| <90.00,<30.00 | |
| MFCD00066650 | |
| 0.90 g/cm3 | |
| 1 L | |
| O | |
| 18.02 | |
| Poly Bottle |
Safety and Handling
H314+H335
P261+P264+P271+P280
Danger
Recommended Storage : Room Temperature