missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 24°C |
| CAS Min % | ≥70.0000%, ≥20.0000% |
| CAS Max % | <90.0000%, <30.0000% |
| Linear Formula | NH4OH |
| UN Number | UN2672 |
| Beilstein | 3587154 |
| Density | 0.9 g/mL at 25 °C(lit.) |
| Quantity | 10 x 1 mL |
| IUPAC Name | azanium;hydroxide |
| Explosion Limit | 0.27 |
| Show More |
Safety and Handling
| Hazard Statement | P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391 |
| ShelfLife | 1080 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Ammonia solutions |
| Recommended Storage | Room Temp |