missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Honeywell Fluka™

ACS Reagent, 28.0-30.0% NH3 basis

Supplier:  Honeywell-Fluka 22122825L

 View more versions of this product

Catalog No. 6000307


Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Ammonium Hydroxide Solution
H5NO
MFCD00066650
3587154
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
35.05
CHEBI:18219
ACS Reagent
≤0.002%
1336-21-6
NH4OH
28 to 30% NH3 basis
0.9g/mL at 25°C
2.5 L
N.O
0.27
14923
35.05g/mol
Glass Bottle

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
N.O
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions