missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Honeywell Fluka™

Catalog No. p-7071891
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000304 100 mL
6000309 500 mL
6000306 1 L
6000307 2.5 L
6000310 6 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000304 Supplier Honeywell Chemicals Supplier No. 221228100ML
Only null left

ACS Reagent, 28.0-30.0% NH3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Linear Formula NH4OH
Concentration 28 to 30% NH3 basis
Beilstein 3587154
Density 0.9g/mL at 25°C
Quantity 100 mL
IUPAC Name azanium;hydroxide
Explosion Limit 0.27
Formula Weight 35.05g/mol
Grade ACS Reagent
Packaging Glass Bottle
Residue after Ignition ≤0.002%
Chemical Name or Material Ammonium Hydroxide Solution
Show More Show Less