missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Reagent, ACS, 28-30%, Spectrum™ Chemical

A1195, 1336-21-6, NH4OH

$226.70 - $6,848.40

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O
View More Specs

Products 14
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600606
View Documents
Spectrum Chemical Mfg Cor
A1195100ML
100 mL Amber Glass Bottle N/A
Only null left
 
18600614
View Documents
Spectrum Chemical Mfg Cor
A1195500MLGL
500 mL Amber Glass Bottle N/A
Only null left
 
18600608
View Documents
Spectrum Chemical Mfg Cor
A119525LTGL
2.5 L Glass Bottle N/A
Only null left
 
18600607
View Documents
Spectrum Chemical Mfg Cor
A119525LTPV
2.5 L Poly-Coated Glass Bottle N/A
Only null left
 
18600615
View Documents
Spectrum Chemical Mfg Cor
A1195500MLGLCS6
6 x 500 mL Amber Glass Bottle N/A
Only null left
 
18600609
View Documents
Spectrum Chemical Mfg Cor
A11954LTPV
4 L Poly-Coated Glass Bottle N/A
Only null left
 
18600611
View Documents
Spectrum Chemical Mfg Cor
A119525LTGLCS4
4 x 2.5 L Glass Bottle N/A
Only null left
 
18600610
View Documents
Spectrum Chemical Mfg Cor
A119525LTPVCS4
4 x 2.5 L Poly-Coated Glass Bottle N/A
Only null left
 
18600618
View Documents
Spectrum Chemical Mfg Cor
A1195135LT
13.5 L Poly Drum N/A
Only null left
 
18600619
View Documents
Spectrum Chemical Mfg Cor
A119525LTGLCS6
6 x 2.5 L Glass Bottle N/A
Only null left
 
18600617
View Documents
Spectrum Chemical Mfg Cor
A119525LTPVCS6
6 x 2.5 L Poly-Coated Glass Bottle N/A
Only null left
 
18600612
View Documents
Spectrum Chemical Mfg Cor
A11954LTPVCS4
4 x 4 L Poly-Coated Glass Bottle N/A
Only null left
 
18600613
View Documents
Spectrum Chemical Mfg Cor
A119520LTPL
20 L Poly Drum N/A
Only null left
 
18600616
View Documents
Spectrum Chemical Mfg Cor
A1195200LTBL
200 L Drum N/A
Only null left
 
Description

Description

Spectrum™ Chemical Ammonium Hydroxide, Reagent, ACS is a solution of amonia in water. Aqueous amonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Also used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent. It is also commmonly used in numerous household cleaning agents, such as window cleaning formula. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Hydroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
N.O
H5NO
MFCD00066650
amine hydrate

Specifications

1336-21-6
H5NO
UN2672
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
28 to 30%
Amber Glass Bottle
100%
MFCD00066650
100 mL
N.O
35.05
ACS
0.002%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'