missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Reagent, ACS, 28-30%, Spectrum™ Chemical

Catalog No. 18600619
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
13.5 L
20 L
4 x 2.5 L
4 x 4 L
6 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600619 6 x 2.5 L Glass Bottle
18600606 100 mL Amber Glass Bottle
18600614 500 mL Amber Glass Bottle
18600608 2.5 L Glass Bottle
18600607 2.5 L Poly-Coated Glass Bottle
18600615 6 x 500 mL Amber Glass Bottle
18600609 4 L Poly-Coated Glass Bottle
18600611 4 x 2.5 L Glass Bottle
18600610 4 x 2.5 L Poly-Coated Glass Bottle
18600618 13.5 L Poly Drum
18600617 6 x 2.5 L Poly-Coated Glass Bottle
18600612 4 x 4 L Poly-Coated Glass Bottle
18600613 20 L Poly Drum
18600616 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 18600619 Supplier Spectrum Chemical Mfg Cor Supplier No. A11952.5LTGLCS6
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1195, 1336-21-6, NH4OH

Spectrum™ Chemical Ammonium Hydroxide, Reagent, ACS is a solution of amonia in water. Aqueous amonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Also used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent. It is also commmonly used in numerous household cleaning agents, such as window cleaning formula. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Hydroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 100%
UN Number UN2672
Quantity 6 x 2.5 L
Percent Purity 28 to 30%
Grade ACS
Packaging Glass Bottle
Residue after Ignition 0.002%