missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, For Phenols, Certified, 0.50N ±0.01N (0.5M), LabChem™

Catalog No. p-5388991
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC110701 500 mL
LC110704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC110701 Supplier LabChem Supplier No. LC110701
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Max % 99.19, 0.81
Linear Formula NH4OH
Concentration 0.50N ±0.01N (0.5M)
Solubility Information Soluble in water
Quantity 500 mL
Formula Weight 35.05
Grade Certified
Identification Passes Test
Odor Ammonia
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, For Phenols
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature