missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, For Hardness, Certified, 19.2 to 20.8% (v/v), LabChem™

$69.27 - $402.13

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC111601
View Documents
LabChem
LC111601
500 mL
Each for $69.27
Only null left
 
LC111602
View Documents
LabChem
LC111602
1 L
Each for $109.33
Only null left
 
LC111604
View Documents
LabChem
LC111604
4 L
Each for $223.47
Only null left
 
LC111606
View Documents
LabChem
LC111606
10 L
Each for $402.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O

Specifications

Colorless
94.95,5.05
NH4OH
0.98g/mL
Soluble in water
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
Certified
Ammonia
0.98g/mL
Liquid
as NH4OH
1336-21-6,7732-18-5
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
500 mL
N.O
35.05
CHEBI:18219
19.2 to 20.8% (v/v)
Passes Test
Poly Bottle
Traceable to NIST
Ammonium Hydroxide, For Hardness
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'