Learn More
Ammonium Hydroxide, For Hardness, Certified, 19.2 to 20.8% (v/v), LabChem™
Supplier: LabChem LC111602
Specifications
| Ammonium Hydroxide | |
| For Hardness | |
| 94.95,5.05 | |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| 1 L | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| 19.2 to 20.8% (v/v) | |
| Passes Test | |
| Poly Bottle | |
| Traceable to NIST |
| Colorless | |
| 1336-21-6,7732-18-5 | |
| as NH4OH | |
| NH4OH | |
| 0.98g/mL | |
| Soluble in water | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| 35.05 | |
| Certified | |
| Ammonia | |
| 0.98g/mL | |
| Liquid |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| 14923 | |
| amine hydrate |
| H5NO | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| CHEBI:18219 | |
| N.O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.
GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature