missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Certified, 9.6 to 10.3% (v/v), LabChem™

Catalog No. LC111801
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC111801 500 mL
LC111802 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC111801 Supplier LabChem Supplier No. LC111801
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Max % 97.5, 2.5
Linear Formula NH4OH
Density 0.987g/mL
Solubility Information Soluble in water
Quantity 500 mL
Formula Weight 35.05
Percent Purity 9.6 to 10.3% (v/v)
Grade Certified
Identification Passes Test
Odor Ammonia
Packaging Poly Bottle
Specific Gravity 0.987g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Assay as NH4OH
Chemical Name or Material Ammonium Hydroxide, as NH4OH
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature