missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Certified, 6.00N ±0.20N (6.0M), LabChem™

Catalog No. p-5388987
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC111201 500 mL
LC111202 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC111201 Supplier LabChem Supplier No. LC111201
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Max % 89.3, 10.7
Linear Formula NH4OH
Concentration 6.00N ±0.20N (6.0M)
UN Number UN2672
Density 0.95g/mL
Solubility Information Soluble in water
Quantity 500 mL
Formula Weight 35.05
Grade Certified
Identification Passes Test
Odor Ammonia
Packaging Poly Bottle
Specific Gravity 0.95g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Keep container tightly closed.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature