missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, Certified, 5.0N ±0.1N (5.0M), LabChem™

Catalog No. p-5388985
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Catalog No. Quantity
LC111101 500 mL
LC111102 1 L
LC111104 4 L
3 options

Catalog No. LC111101

Supplier: LabChem LC111101

Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Max % 91.15, 8.85
Linear Formula NH4OH
Concentration 5.0N ±0.1N (5.0M)
Density 0.96g/mL
Solubility Information Soluble in water
Quantity 500 mL
Formula Weight 35.05
Grade Certified
Identification Passes Test
Odor Ammonia
Packaging Poly Bottle
Specific Gravity 0.96g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Keep container tightly closed.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature