missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, Certified, 16.1 to 17.3% (v/v), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 95.8, 4.2 |
| Linear Formula | NH4OH |
| Density | 0.98g/mL |
| Solubility Information | Soluble in water |
| Quantity | 500 mL |
| Formula Weight | 35.05 |
| Percent Purity | 16.1 to 17.3% (v/v) |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If on skin: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. Take off contaminated clothing and wash before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |