missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, for analysis, 20% solution in water

Catalog No. p-4600135
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390070010 1 L Glass bottle
AC390070025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC390070010 Supplier Thermo Scientific Chemicals Supplier No. 390070010

CAS: 1336-21-6 | H5NO | 35.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Color Brown-Green to Brown-Purple
Linear Formula NH4OH
Merck Index 15, 491
Density 0.9200g/mL
Solubility Information Solubility in water: soluble.
Quantity 1 L
IUPAC Name amine hydrate
Formula Weight 35.05
Percent Purity ≥20% (Acidimetry)
Grade Analytical
Packaging Glass bottle
Specific Gravity 0.92
Physical Form Solution
Chemical Name or Material Ammonium hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage
May cause respiratory irritation
Precautionary Statement GHS H Statement:
Causes severe skin burns and eye damage
May cause respiratory irritation
IF SWALLOWED: rinse mouth. Do NOT induce vomiting
Wear protective gloves/protective clothing/eye protection/face protection
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Avoid breathing dust/fume/gas/mist/vapors/spray

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-647-6

RUO – Research Use Only