missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3251282
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABA0020500 500 mL Glass Bottle
RABA002025D 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RABA0020500 Supplier Ricca Chemical Company Supplier No. RABA0020500D1
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Concentration or Composition Notes 28.0-30.0% as NH3
Quantity 500 mL
Percent Purity 28 to 30%
Grade ACS
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide