missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, ACS Reagent (Aqua Ammonia), Reagents

Catalog No. p-200020970
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
194 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Drum
Safety Coated Glass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336794 194 L Poly Drum
01336793 2.5 L Safety Coated Glass
01336792 2.5 L Glass Bottle
01336791 20 L HDPE Winpak
01336790 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336794 Supplier Labchem Inc Supplier No. C5103500194Y
Only null left

Ammonium Hydroxide, ACS Reagent (Aqua Ammonia), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH 13.6
Color Colorless
Boiling Point 36°C
Quantity 194 L
Grade ACS
Packaging Poly Drum
Specific Gravity 0.9
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, ACS Reagent (Aqua Ammonia)