missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, ACS Reagent (Aqua Ammonia), Reagents
Specifications
Specifications
| pH | 13.6 |
| Color | Colorless |
| Boiling Point | 36°C |
| Quantity | 2.5 L |
| Grade | ACS |
| Packaging | Safety Coated Glass |
| Specific Gravity | 0.9 |
| Physical Form | Liquid |
| Chemical Name or Material | Ammonium Hydroxide, ACS Reagent (Aqua Ammonia) |