missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, ACS Grade, 28.0 to 30.0%, LabChem™

Catalog No. LC1105036
Change view
Click to view available options
Quantity:
6 x 2.5 L
Packaging:
HDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC1105036 6 x 2.5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC1105036 Supplier LabChem Supplier No. LC1105036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Boiling Point 27°C
CAS Max % 71, 29
Linear Formula NH4OH
UN Number UN2672
Density 0.89
Solubility Information Soluble in water
Quantity 6 x 2.5 L
Formula Weight 35.05
Percent Purity 28.0 to 30.0%
Grade ACS
Identification Passes Test
Odor Ammonia
Packaging HDPE Bottle
Specific Gravity 0.89
Physical Form Liquid
Assay as NH3
Chemical Name or Material Ammonium Hydroxide, as NH3
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 215-647-6
Recommended Storage Room Temperature