missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N
CAS: 1336-21-6 | H5NO | 35.05 g/mol
$97.26 - $457.22
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| IUPAC Name | azanium;hydroxide |
| SMILES | N.O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| CHEBI:18219 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| 14923 | |
| azanium;hydroxide |
Specifications
| 1336-21-6 | |
| 0.1N | |
| H5NO | |
| Colorless | |
| N.O | |
| 35.05 | |
| 14923 | |
| 35.05 |
| Ammonia-like | |
| Liquid | |
| MFCD00066650 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| azanium;hydroxide | |
| 1 Unit | |
| CHEBI:18219 | |
| Ammonium hydroxide |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 215-647-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only