missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 6 Normal, Reagents

Catalog No. 01335994
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335994 1 L
01335993 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335994 Supplier Labchem Inc Supplier No. CA0206001A
Only null left

Ammonium Hydroxide, 6 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 36°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9567
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 6 Normal