missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 6.0 N Solution, Spectrum™ Chemical

Catalog No. 18600605
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600605 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18600605 Supplier Spectrum Chemical Mfg Cor Supplier No. A5401LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A-540, 1336-21-6

Spectrum™ Chemical Ammonium Hydroxide, 6.0 N Solution , is a solution of ammonia in water. Aqueous ammonia, as it's otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Used to dissolve silver oxide residues, such as the ones formed from Tollens' reagent, it is also commonly used in numerous cleaning agents, such as window cleaning formulas as well as a complexant that is manufactured to parts-per-billion trace impurities and has been used in the semiconductor industry to dissolve heavy or precious metal components.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 6%, 92%
Physical Form Liquid
Quantity 1 L
UN Number UN2672
Concentration or Composition (by Analyte or Components) 6.0 N +/-0.2
Packaging Poly Bottle
Chemical Name or Material Ammonium Hydroxide,