missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 50% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company R064000020F

 View more versions of this product

Catalog No. R064000020F


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Hydroxide
1336-21-6,7732-18-5
51.0,51.0
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
48.5 to 51.5% v/v
Cubitainer
Liquid
Colorless
49.0,49.0
H5NO
50% (v/v)
20 L
N.O
35.05
CHEBI:18219
Laboratory
Aqueous Solution

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
CHEBI:18219
N.O
H5NO
MFCD00066650
14923
amine hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents

Promotions

Promotions