missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 50% (v/v), Ricca Chemical
$130.40 - $631.53
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| IUPAC Name | amine hydrate |
| SMILES | N.O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
640-16
|
Ricca Chemical Company
64016 |
500 mL | Natural Poly Bottle |
Each for $130.40
|
|
||||
|
640-32
|
Ricca Chemical Company
64032 |
1 L | Natural Poly Bottle |
Each for $199.70
|
|
||||
|
640-1
|
Ricca Chemical Company
RI6401 |
4 L | Natural Poly Bottle |
Each for $409.30
|
|
||||
|
R064000020F
|
Ricca Chemical Company
R064000020F |
20 L | Cubitainer |
Each for $631.53
|
|
||||
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| CHEBI:18219 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| 14923 | |
| amine hydrate |
Specifications
| Colorless | |
| 49.0,49.0 | |
| H5NO | |
| 50% (v/v) | |
| 500 mL | |
| N.O | |
| 35.05 | |
| CHEBI:18219 | |
| Laboratory | |
| Aqueous Solution | |
| Ammonium Hydroxide |
| 1336-21-6,7732-18-5 | |
| 51.0,51.0 | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 14923 | |
| 48.5 to 51.5% v/v | |
| Natural Poly Bottle | |
| Liquid |
Safety and Handling
ShelfLife : 24 months