missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 29% v/v, Reagents

Catalog No. 01336116
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01336116 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336116 Supplier Labchem Inc Supplier No. CA0074004A
Only null left

Ammonium Hydroxide, 29% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 36°C
Quantity 4 L
Percent Purity 29%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.971
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 29% v/v