missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 29%, Electronic/Cleanroom Grade, 28.0-30.0%, Spectrum™ Chemical

Catalog No. 18608676
Change view
Click to view available options
Quantity:
1 gal.
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608676 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18608676 Supplier Spectrum Chemical Mfg Cor Supplier No. A11871GL
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7664-41-7
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 1
Quantity 1 gal.
Percent Purity 28 to 30%
Grade Electronic/Cleanroom
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.