missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 26 DEG Be, Technical, Approx. 29%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Concentration | 26 DEG Be |
| Quantity | 4 L |
| Percent Purity | ∼29% |
| Grade | Technical |
| Packaging | Poly-Coated Glass Bottle |
| Chemical Name or Material | Ammonium Hydroxide |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.