missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 26 DEG Be, Technical, Approx. 29%, Spectrum™ Chemical

Catalog No. 18608675
Change view
Click to view available options
Quantity:
4 L
20 L
55 gal.
Packaging:
Poly Drum
Poly-Coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608675 4 L Poly-Coated Glass Bottle
18608673 20 L Poly Drum
18608674 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608675 Supplier Spectrum Chemical Mfg Cor Supplier No. A12004LTPV
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7664-41-7

Specifications

CAS Max % 1
Concentration 26 DEG Be
Quantity 4 L
Percent Purity ∼29%
Grade Technical
Packaging Poly-Coated Glass Bottle
Chemical Name or Material Ammonium Hydroxide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.