missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 26° Baume’ (Aqua Ammonia), Reagents

Catalog No. 01336798
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
15 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336798 15 gal.
01336797 2.5 L
01336796 4 L
01336795 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336798 Supplier Labchem Inc Supplier No. C510300015E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 26° Baume’ (Aqua Ammonia), Technical, for general laboratory use. Store below 30°C. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH 13.6
Color Colorless
Boiling Point 36°C
Quantity 15 gal.
Packaging Poly Drum
Specific Gravity 0.9
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 26° Baume’ (Aqua Ammonia)