missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 25% v/v, Reagents

Catalog No. 01335976
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335976 1 L
01335975 20 L
01335974 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335976 Supplier Labchem Inc Supplier No. CA1003001A

Ammonium Hydroxide, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 36°C
Quantity 1 L
Percent Purity 25%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.975
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 25% v/v