missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 20% v/v, Reagents

Catalog No. 01336022
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336022 20 L
01336024 10 L
01336023 1 L
01336021 4 L
01336020 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336022 Supplier Labchem Inc Supplier No. CA01530020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 20% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 47°C
Quantity 20 L
Percent Purity 20%
Grade Reagent
Packaging Cubitainer
Specific Gravity 0.98
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 20% v/v