missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 2.00 N, Ricca Chemical

Catalog No. R06430004A
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R06430004A 4 L Natural Poly Bottle
64316 500 mL Natural Poly Bottle
64332 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R06430004A Supplier Ricca Chemical Company Supplier No. R06430004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Min % 87.0, 10.83
CAS Max % 91.0, 11.27
Concentration 1.999 to 2.001 N
Synonym ammonium hydroxide
Quantity 4 L
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Solution
Assay Normal at 20°C
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
ShelfLife 24 months