missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 2.00 N, Ricca Chemical

Catalog No. 64316
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
64316 500 mL Natural Poly Bottle
64332 1 L Natural Poly Bottle
R06430004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 64316 Supplier Ricca Chemical Company Supplier No. 64316
Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Min % 87.0, 10.83
CAS Max % 91.0, 11.27
Concentration 1.999 to 2.001 N
Concentration or Composition Notes 2.00 N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Quantity 500 mL
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Solution
Assay Normal at 20°C
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
ShelfLife 24 months