missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 16.7% v/v, Electronic Grade, Reagents

Catalog No. 01336027
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
55 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336027 4 L
01336029 1 L
01336028 20 L
01336026 500 mL
01336025 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336027 Supplier Labchem Inc Supplier No. CA0152004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 16.7% v/v, Electronic Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 60°C
Quantity 4 L
Percent Purity 16.7%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.982
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 16.7% v/v, Electronic Grade