missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 15% w/w in Ethanol, Reagents

Catalog No. 01336080
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336080 4 L
01336081 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336080 Supplier Labchem Inc Supplier No. CA0112004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 15% w/w in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 1336-21-6
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Quantity 4 L
Percent Purity 15%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8041
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 15% w/w in Ethanol