missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 10% (w/w) as NH?, Reagents
Ammonium Hydroxide, 10% (w/w) as NH?, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CA01980020F
Specifications
| >11.5 | |
| Colorless | |
| 7732-18-5,1336-21-6 | |
| Aqua Ammonia | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 10% | |
| Cubitainer | |
| Liquid |
| Ammonium Hydroxide, 10% (w/w) as NH | |
| 70°C | |
| H2O | |
| 20 L | |
| O | |
| 18.02 | |
| Reagent | |
| 0.9631 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Aqua Ammonia | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |