missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 10% (w/w) as NH?, Reagents

Catalog No. 01335998
Change view
Click to view available options
Quantity:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335998 10 L
01335997 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335998 Supplier Labchem Inc Supplier No. CA01980010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 10% (w/w) as NH?, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 70°C
Quantity 10 L
Percent Purity 10%
Grade Reagent
Packaging Cubitainer
Specific Gravity 0.9631
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 10% (w/w) as NH