missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 10% w/v, Reagents
Ammonium Hydroxide, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C71035204A
Spécifications
| >11.5 | |
| Colorless | |
| 7732-18-5,1336-21-6 | |
| Aqua Ammonia | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 10% w/v | |
| Poly Bottle | |
| Liquid |
| Ammonium Hydroxide, 10% w/v | |
| 69°C | |
| H2O | |
| 4 L | |
| O | |
| 18.02 | |
| Reagent | |
| 0.987 |
Identifiants chimiques
| 7732-18-5 | |
| 18.02 | |
| Aqua Ammonia | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |