missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 10% v/v, Reagents

Catalog No. p-200021446
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Hedpak
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336078 1 L Poly Bottle
01336077 20 L Hedpak
01336076 20 L Cubitainer
01336075 4 L Poly Bottle
01336074 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336078 Supplier Labchem Inc Supplier No. CA0115001A
Only null left

Ammonium Hydroxide, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 69°C
Quantity 1 L
Percent Purity 10%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9892
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 10% v/v