missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 10.0 Normal, Reagents

Catalog No. 01336030
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336030 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336030 Supplier Labchem Inc Supplier No. CA015100500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonium Hydroxide, 10.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 36°C
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9299
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 10.0 Normal