missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 1:1, 50% v/v, Reagents

Catalog No. 01335981
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
15 gal.
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335981 15 gal.
01335980 1 L
01335979 200 L
01335978 20 L
01335977 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335981 Supplier Labchem Inc Supplier No. CA10020015E

Ammonium Hydroxide, 1:1, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH >11.5
Color Colorless
Boiling Point 36°C
Quantity 15 gal.
Percent Purity 50%
Grade Reagent
Packaging Poly Drum
Specific Gravity 0.95
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, 1:1, 50% v/v