missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, 1.0N Standardized Solution

Catalog No. AA35614K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35614K2 1 L
AA35614K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35614K2 Supplier Thermo Scientific Chemicals Supplier No. 035614K2

CAS: 1336-21-6 | H5NO | 35.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Odor Ammonia-like
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
IUPAC Name amine hydrate
Quantity 1 L
Formula Weight 35.05
Chemical Name or Material Ammonium hydroxide
Hazard Category H315-H318
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
EINECSNumber 215-647-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only