missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide, 1.00 N, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 92.0, 5.73 |
| CAS Max % | 96.0, 5.96 |
| Concentration or Composition Notes | 1.00Normal |
| Quantity | 500 mL |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Chemical Name or Material | Ammonium Hydroxide |